Sign In to View Organizational & Contract Pricing.
About This Item
Empirical Formula (Hill Notation):
C15H24N2O2
CAS Number:
Molecular Weight:
264.36
NACRES:
NA.25
PubChem Substance ID:
UNSPSC Code:
12352200
MDL number:
assay
≥98% (HPLC)
Quality Level
form
powder
solubility
methanol: 10 mg/mL, clear, colorless
storage temp.
2-8°C
SMILES string
[O-][N+]12CCCC3CN4C(CCCC4=O)C(CCC1)C23
InChI
1S/C15H24N2O2/c18-14-7-1-6-13-12-5-3-9-17(19)8-2-4-11(15(12)17)10-16(13)14/h11-13,15H,1-10H2/t11-,12+,13+,15-,17?/m0/s1
InChI key
XVPBINOPNYFXID-LHDUFFHYSA-N
Biochem/physiol Actions
Alkaloid isolated from Sophora flavescens. Antifribotic. Neuroprotective. Traditional chinese medicine used in the treatment against hepatitis B virus.
Still not finding the right product?
Explore all of our products under Oxymatrine
Storage Class
11 - Combustible Solids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Ruiying Wang et al.
Stem cell research & therapy, 10(1), 20-20 (2019-01-13)
Cardiotoxicity remains an important concern in drug discovery and clinical medication. Meanwhile, Sophora tonkinensis Gapnep. (S. tonkinensis) held great value in the clinical application of traditional Chinese medicine, but cardiotoxic effects were reported, with matrine, oxymatrine, cytisine, and sophocarpine being
Man Huang et al.
Journal of ethnopharmacology, 143(1), 228-235 (2012-07-04)
Oxymatrine is extracted from the traditional Chinese herb Sophora flavescens Ait, possesses anti-inflammatory, anti-oxidative and anti-apoptotic properties, and has been used for the treatment of chronic viral hepatitis and many other diseases. This study aimed to investigate the effects of
Xiaopei Yuan et al.
Journal of ethnopharmacology, 137(3), 1477-1485 (2011-09-01)
The combination of Radix Angelicae sinensis (Oliv.) Diels and Radix Sophora flavescens Ait. was extensively used in traditional Chinese medicine to treat inflammatory diseases, such as acne, heart disease, and hepatitis. Sodium ferulate (SF) and oxymatrine (OMT) were effective component
Global Trade Item Number
| SKU | GTIN |
|---|---|
| O0891-10MG | 04061832402000 |