Skip to Content
Merck

Skip To

BDPH9BCC3CD7

Bide Pharmatech

7-Bromo-2,4-dichlorothieno[3,2-d]pyrimidine

Synonym(s):

7-bromo-2,4-dichlorothieno[3,2-d]pyrimidine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C6H1N2Cl2Br1S1
CAS Number:
Molecular Weight:
283.96
MDL number:
Product BDPH9BCC3CD7 is not currently sold in your country.


Inchi Code

1S/C6HBrCl2N2S/c7-2-1-12-4-3(2)10-6(9)11-5(4)8/h1H

Inchi Key

SELNIRJYLYMJKG-UHFFFAOYSA-N

Shipping Temperature

Shipped in cold pack

Purity

95%

Storage temp.

Keep in dark place, inert atmosphere, store in freezer, under -20C

Country of Origin

CN

Physical Form

Pale-yellow to Yellow-brown Solid



Still not finding the right product?


signalword

Warning

pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library