Skip to Content
Merck

Skip To

BDPH9BCDFD1A

Bide Pharmatech

3-Amino-2-methylbutan-2-ol

Synonym(s):

3-amino-2-methyl-2-butanol

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C5H13O1N1
CAS Number:
Molecular Weight:
103.16
MDL number:
Product BDPH9BCDFD1A is not currently sold in your country.


Inchi Code

1S/C5H13NO/c1-4(6)5(2,3)7/h4,7H,6H2,1-3H3

Inchi Key

OVKDLPZRDQTOJW-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

95%

Storage temp.

Keep in dark place, inert atmosphere, room temperature

Country of Origin

CN

Physical Form

Solid or semi-solid or liquid or lump



Still not finding the right product?

Explore all of our products under 3-Amino-2-methylbutan-2-ol



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library