Skip to Content
Merck

Skip To

BDPH9BCDFD24

Bide Pharmatech

Ethyl 1-bromo-2-naphthoate

Synonym(s):

ethyl 1-bromo-2-naphthoate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C13H11O2Br1
CAS Number:
Molecular Weight:
279.13
MDL number:
Product BDPH9BCDFD24 is not currently sold in your country.


Inchi Code

1S/C13H11BrO2/c1-2-16-13(15)11-8-7-9-5-3-4-6-10(9)12(11)14/h3-8H,2H2,1H3

Inchi Key

TXDQMAGPAKABJU-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

98%

Storage temp.

Sealed in dry, 2-8C

Country of Origin

CN

Physical Form

Solid or liquid



Still not finding the right product?

Explore all of our products under Ethyl 1-bromo-2-naphthoate


hstatements

signalword

Warning

pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library