Skip to Content
Merck

Skip To

BDPH9C021A0F

Bide Pharmatech

1-(a-D-ribofuranosyl)uracil

Synonym(s):

1-((2S,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)pyrimidine-2,4(1H,3H)-dione

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C9H12O6N2
CAS Number:
Molecular Weight:
244.2
MDL number:
Product BDPH9C021A0F is not currently sold in your country.


Inchi Code

1S/C9H12N2O6/c12-3-4-6(14)7(15)8(17-4)11-2-1-5(13)10-9(11)16/h1-2,4,6-8,12,14-15H,3H2,(H,10,13,16)/t4-,6-,7-,8+/m1/s1

Inchi Key

DRTQHJPVMGBUCF-JBBNEOJLSA-N

Shipping Temperature

Normal

Purity

95%

Storage temp.

Sealed in dry, 2-8C

Country of Origin

CN

Physical Form

White to off-white solid



Still not finding the right product?

Explore all of our products under 1-(a-D-ribofuranosyl)uracil


hstatements

signalword

Warning

pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library