Merck
ReCycler® Solvent Program
Our high-quality standards apply not only to solvents themselves but also to the packaging they are supplied in. Discover our ReCycler® Solvent Program, an initiative aimed at reducing packaging waste and promoting sustainability.
Each product is carefully developed, tested, and matched to its specifications. The program provides customers working with solvents with reusable containers, instead of single-use packaging, for the safe transport and storage of solvents. This helps to decrease the amount of waste generated and reduces the need for frequent container replacements. By participating in the program, customers can not only help the environment but also benefit from cost savings and improved operational efficiency.
For checking the availability of this program in your country, please get in touch with your local Customer Service.
For any question about your returnable container project please contact us:
Please note that whereas the returnable drums are available in various countries (incl. Western Europe), the returnable barrels are only available in these countries: Austria, Belgium, Denmark, Finland, France, Germany, Italy, Ireland, Luxembourg, Netherlands, Norway, Portugal, Spain, Sweden, Switzerland, and the UK
Returnable solvent containers
Our returnable containers are made of high-quality stainless steel, which allows them to withstand multiple uses and protect the solvents from contamination. They are available in:
Returnable stainless-steel barrels in 10 L, 30 L, and 185 L volumes
Stainless-steel barrels
  • Stainless steel barrels of 10 L, 30 L, and 185 L sizes. Their return is mandatory and is handled with a deposit system, which is refunded when the barrel is sent back. The stainless-steel barrels are exclusively available in Western Europe. To return empty barrels, please contact your local Customer Service.
Order high-purity solvents in stainless steel barrels
Stainless-steel barrels in 10 L, 25 L, and 190 L sizes
Stainless-steel drums
  • Stainless steel drums of 10 L, 25 L, and 190 L volumes and their return is optional. These are available globally and do not require a deposit. If you are based in Western Europe and would like to return the emptied drums, please contact your local Customer Service.
Order classical solvents in stainless-steel drums
Solvents
The following solvent grades are available in the ReCycler® Solvent Program, either in stainless steel barrels or drums.
  • LiChrosolv® solvents for liquid chromatography
  • SupraSolv® solvents for gas chromatography
  • Uvasol® solvents for UV/Vis and infrared spectroscopy
  • Prepsolv® solvents for preparative chromatography
  • EMSURE® solvents for advanced and regulated analyses
  • EMPARTA® solvents for routine analytical work
  • EMPLURA® solvents for preparative laboratory work
Features & Benefits
  • High-quality and inert stainless steel containers maintain the quality of the solvent
  • Strong and secure packaging material maximizes user safety and minimizes environmental pollution risks
  • An optimum removal of any residual quantities by Merck withdrawal systems
  • Merck withdrawal systems help avoid contact with hazardous solvents and prevent the emission of solvent vapors into the environment
  • Returnable stainless steel solvent containers reduce packaging waste and save raw materials
How does the ReCycler® Solvent Program Work?
1. Validated barrel cleaning
Validated barrel cleaning
Production / Filling at Merck KGaA, Darmstadt, Germany.
    2. Customer Order
    Delivery truck
    Transportation to the customer site and consumption.
      3. Pick-up & Return
      Company representative
      Please contact your local Merck representative for pick up of emptied container for its return to Merck KGaA, Darmstadt, Germany.
        4. Deposit Credit
        Returnable stainless-steel barrels in 10 L, 30 L, and 185 L volumes
        For stainless-steel barrels only.
          Returnable Solvent Containers Specifications Overview
          Stainless-steel Barrels and Drums, 1. 10 L stainless-steel barrel, 2. 30 L stainless-steel barrel, 3. 185 L stainless-steel barrel, 4. 10 L Stainless Steel Drum, 5. 25 L Stainless Steel Drum, 6. 190 L Stainless Steel Drum
          Stainless-steel Barrels and Drums, 1. 10 L stainless-steel barrel, 2. 30 L stainless-steel barrel, 3. 185 L stainless-steel barrel, 4. 10 L Stainless Steel Drum, 5. 25 L Stainless Steel Drum, 6. 190 L Stainless Steel Drum
          Solvent Quality GradeLiChrosolv® Solvents
          Prepsolv® Solvents
          SupraSolv® Solvents
           EMSURE® Solvents
          EMPARTA® Solvents
          EMPLURA® Solvents
           10 L Stainless Steel Barrel30 L Stainless Steel Barrel185 L Stainless Steel Barrel 10 L Stainless Steel Drum25 L Stainless Steel Drum190 L Stainless Steel Drum
          ReturnabilityMandatoryMandatoryMandatory OptionalOptionalOptional
          DepositYesYesYes NoNoNo
          AvailabilityWestern EuropeWestern EuropeWestern Europe GlobalGlobalGlobal
          Height31 cm44 cm97 cm 35 cm52 cm88 cm
          Diameter28 cm37 cm58 cm 24 cm29 cm59.5 cm
          Volume12 L33.5 L206 L 12 L28 L215 L
          Filling quantity10 L30 L185 L 10 L25 L190 L
          Weight (empty)5.5 kg9.6 kg31 kg 1.9 kg3.8 kg18 kg
          Recommended working pressure0.2 bar0.2 bar0.2 bar 0.2 bar0.2 bar0.2 bar
          max. working pressure0.5 bar0.5 bar0.5 bar 0.3 bar0.3 bar0.3 bar 
          Number per pallet1162 15112
          Openings2" centrally and 3/4" decentrally located2" centrally and 3/4" decentrally located2" centrally and 3/4" decentrally located 2" decentrally located2" decentrally located2" decentrally located and 3/4" decentrally located
          MaterialStainless steel 1.4301Stainless steel 1.4301Stainless steel 1.4301 Stainless steel 1.4301Stainless steel 1.4301Stainless steel 1.4301
          Accessories       
          Opening key for barrels & drums1.08803.00011.08803.00011.08803.0001 1.08803.00011.08803.00011.08803.0001
          Antistatic set
          (mandatory for Option 1 and Option 2)
          1.07070.00011.07070.00011.07070.0001 1.07070.00011.07070.00011.07070.0001
          Option 1: Manual Withdrawal System1.01123.00011.01123.00011.19171.0001 1.01114.00011.01114.00011.19171.0001
          Option 2: Inert-gas system
          Withdrawal System + dip tube
          1.06710.0001
          9.67100.1040
          1.06710.0001
          9.67100.1041
          1.06710.0001
          9.67100.1185
           1.06710.0001
          9.67100.1010
          1.06710.0001
          9.67100.1025
          1.06710.0001
          9.67100.1190
          Comprehensive Solvent Management Solutions
          With an extensive portfolio of solvents, containers, withdrawal systems, and a multitude of accessories, we deliver total solvent management solutions for your individual applications. All components are fully compatible to permit simple, contamination-free, and safe handling of solvents and their withdrawal, to ensure maximum protection of your lab personnel and the environment.
          Explore our range of withdrawal systems and safety accessories
          Related Products
          Product No.Description
          100030Acetonitrile gradient grade for liquid chromatography LiChrosolv® Reag. Ph Eur
          106007Methanol gradient grade for liquid chromatography LiChrosolv® Reag. Ph Eur
          100029Acetonitrile hypergrade for LC-MS LiChrosolv® solvents
          106035Methanol hypergrade for LC-MS LiChrosolv® solvents
          114291Acetonitrile isocratic grade for liquid chromatography LiChrosolv® solvents
          106018Methanol for liquid chromatography LiChrosolv® solvents
          104371n-Hexane for gas chromatography ECD and FID SupraSolv® solvents
          100983Ethanol absolute for analysis EMSURE® ACS,ISO,Reag. Ph Eur
          106009Methanol for analysis EMSURE® ACS,ISO,Reag. Ph Eur
          100014Acetone for analysis EMSURE® ACS,ISO,Reag. Ph Eur
          102445Chloroform for analysis EMSURE® ACS,ISO,Reag. Ph Eur
          1096342-Propanol for analysis EMSURE® ACS,ISO,Reag. Ph Eur
          109623Ethyl acetate for analysis EMSURE® ACS,ISO,Reag. Ph Eur
          113358Acetonitrile for preparative chromatography Prepsolv™ solvents
          113351Methanol for preparative chromatography Prepsolv™ solvents
          104391n-Hexane for liquid chromatography LiChrosolv®
          1010402-Propanol gradient grade for liquid chromatography LiChrosolv®
          108101Tetrahydrofuran for liquid chromatography LiChrosolv®
          111727Ethanol gradient grade for liquid chromatography LiChrosolv®
          115333Water for chromatography (LC-MS Grade) LiChrosolv®
          Related Resources