Produktname
D-Cycloserin synthetic
Produktnummer
c3909
Produktmarke
sigma
CAS-Nummer
68-41-7
Molekulargewicht
102.09
Test | Spezifikation |
|---|---|
| APPEARANCE | WHITE TO OFF-WHITE POWDER |
| SOLUBILITY | CLEAR COLORLESS TO LIGHT YELLOW OR LIGHT PINK SOLUTION AT 50MG/ML IN WATER |
| CARBON | 34.6 TO 36.0% |
| NITROGEN | 26.4 TO 27.9% |
| SPECIFIC ROTATION | +110 TO +120DEG (C=1 IN WATER) |
| PURITY BY THIN LAYER CHROMATOGRAPHY | NLT 98% |
| DOCUMENT # C3909/08/11/99/3 |