Fortfahren mit
A5259
Arecaidin -hydrochlorid
analytical standard
Synonym(e):
1,2,5,6-Tetrahydro-1-methylnicotinsäure -hydrochlorid
Anmelden zur Ansicht der Organisations- und Vertragspreise.
Über diesen Artikel
Empirische Formel (Hill-System):
C7H11NO2 · HCl
CAS-Nummer:
Molekulargewicht:
177.63
UNSPSC Code:
41116107
PubChem Substance ID:
MDL number:
grade
analytical standard
technique(s)
HPLC: suitable, gas chromatography (GC): suitable
application(s)
food and beverages
forensics and toxicology
veterinary
format
neat
storage temp.
−20°C
SMILES string
Cl.CN1CCC=C(C1)C(O)=O
Application
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
signalword
Warning
pictograms
Exclamation mark
hcodes
Hazard Classifications
Acute Tox. 4 Oral
Lagerklasse
11 - Combustible Solids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)