Product Name
Poly(ethylene glycol) BioUltra, 600
Product Number
87333
Product Brand
sigma
CAS Number
25322-68-3
Molecular Weight
Test
Specification
APPEARANCE (COLOR)Colorless or White
APPEARANCE (FORM)Liquid or Viscous Liquid or Paste
Molecular Number Mn (GPC-Analysis)Report Result
Molecular Weight Mp (GPC-Analysis)Report Result
Molecular Weight Mw (GPC-Analysis)Report Result
Molecular Weight Mz (GPC-Analysis)Report Result
Polydispersity Index (PDI) (GPC-Analysis)Report Result
DENSITY D20/41.115 - 1.144
REFRACTIVE INDEX N20/D1.468 - 1.470
WATER≤ 1.0 %
SULFATED ASH≤ 0.20 %
INFRARED SPECTRUMCONFORMS TO STRUCTURE
VISCOSITY (ROTATION)150 - 190 mPas
VISCOSITY (CONDITIONS)NEAT, 20 C
HYDROXYL VALUE178 - 197 mg KOH/g
TRACES PEROXIDE (AS H2O2)≤ 20.00 mg/kg
METAL TRACE ANALYSIS (ICP)CORRESPONDS TO REQUIREMENTS
ALUMINIUM (ICP)≤ 5 mg/kg
BARIUM (ICP)≤ 5 mg/kg
BISMUTH (ICP)≤ 5 mg/kg
CALCIUM (ICP)≤ 10 mg/kg
CADMIUM (ICP)≤ 5 mg/kg
COBALT (ICP)≤ 5 mg/kg
CHROMIUM (ICP)≤ 5 mg/kg
COPPER (ICP)≤ 5 mg/kg
IRON (ICP)≤ 5 mg/kg
POTASSIUM (ICP)≤ 500 mg/kg
LITHIUM (ICP)≤ 5 mg/kg
MAGNESIUM (ICP)≤ 5 mg/kg
MANGANESE (ICP)≤ 5 mg/kg
MOLYBDENUM (ICP)≤ 5 mg/kg
SODIUM (ICP)≤ 200 mg/kg
NICKEL (ICP)≤ 5 mg/kg
LEAD (ICP)≤ 5 mg/kg
STRONTIUM (ICP)≤ 5 mg/kg
ZINC (ICP)≤ 5 mg/kg
ARSENIC TRACES (MHS-AAS)≤ 0.10 mg/kg
TOTAL SULFUR AS SO4 (ICP)≤ 50 mg/kg
CHLORIDE (CL)≤ 50 mg/kg
SULFATE (SO4)≤ 50 mg/kg
SOLUBILITY (METHOD)1.500 G IN 30 ML H2O
APPEARANCE (SOLUTION)CLEAR COLORLESS
PH (SOLUTION)5.5 - 7.0
RESIDUE (FILTER TEST)NO RESIDUE
UV - ABS. AT 260 NM≤ 0.060
UV - ABS. AT 280 NM≤ 0.030
RemarkMOLECULAR MASS(CALCULATED FROM HYDROXYL VALUE): 570 - 630
RECOMMENDED RETEST PERIOD42 MONTHS