Product Name
Dexamethasone powder, BioReagent, suitable for cell culture, ≥97%
Product Number
d4902
Product Brand
sigma
CAS Number
50-02-2
Molecular Weight
392.46
Test
Specification
APPEARANCEWHITE TO OFF-WHITE POWDER
SOLUBILITYCLEAR COLORLESS SOLUTION AT 25MG/ML IN METHANOL
UV-VIS SPECTRUMEMM = 15.0 TO 15.5 AT LAMBDA MAX 239 TO 241NM IN METHANOL
PURITY BY HPLCNLT 98%
LOSS ON DRYINGNMT 0.5%
CELL CULTURE TESTPASS
RECOMMENDED RETEST5 YEARS
NOTEDOCUMENT # D4902:6/01-1