Product Name
TERGITOL Type NP-40
Product Number
np40
Product Brand
sigma
CAS Number
127087-87-0
Molecular Weight
Test
Specification
APPEARANCEWHITE POWDER
SOLUBILITYCLEAR TO SLIGHTLY HAZY COLORLESS AT 10% AQUEOUS SOLUTION
WATER CONTENTMAXIMUM 1.0%
PH TEST4.0 TO 8.0 (10% AQUEOUS SOLUTION)
NOTETERGITOL IS A REGISTERED TRADEMARK OF UNION CARBIDE CHEMICALS AND PLASTICS CO., INC.
SHELF LIFE2 YEARS
DOCUMENT # T7631/2/29/96/1