Skip to Content
MilliporeSigma

Skip To

856487

Poly(vinylpolypyrrolidone)

cross-linked, powder

Synonym(s):

PVPP, Polyvinylpyrrolidone, cross-linked

Sign In to View Organizational & Contract Pricing.

About This Item

CAS Number:
UNSPSC Code:
12162002
MDL number:


form

powder

crosslinking

cross-linked

mp

>300 °C

SMILES string

C=CN1CCCC1=O

InChI

1S/C6H9NO/c1-2-7-5-3-4-6(7)8/h2H,1,3-5H2

InChI key

WHNWPMSKXPGLAX-UHFFFAOYSA-N

Features and Benefits

Complexes with phenolics and alkaloids for their removal from plant samples, thus preventing their modification of proteins and any interference they may cause in spectrophotometric determinations of protein content. This is also reported to improve stability of enzymes.


Still not finding the right product?

Explore all of our products under Poly(vinylpolypyrrolidone)




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library