Skip to Content
MilliporeSigma

Skip To

BDPH9BCC1DFD

Bide Pharmatech

Ethyl 2-(2-chloropyridin-4-yl)acetate

Synonym(s):

ethyl 2-(2-chloropyridin-4-yl)acetate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Linear Formula:
C9H10O2N1Cl1
CAS Number:
Molecular Weight:
199.64
MDL number:
Product BDPH9BCC1DFD is currently not available for purchase. Contact Technical Service


Inchi Code

1S/C9H10ClNO2/c1-2-13-9(12)6-7-3-4-11-8(10)5-7/h3-5H,2,6H2,1H3

Inchi Key

MFDZGDBPQSJAIC-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

95%

Storage temp.

Inert atmosphere, 2-8C

Country of Origin

CN

Physical Form

Liquid



Still not finding the right product?



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library