Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Pack Size | SKU | Availability | Price |
|---|
About This Item
Product BDPH9BCC2BFB is currently not available for purchase. Contact Technical Service
Inchi Code
1S/C8H7ClN2/c1-11-5-6-3-2-4-7(9)8(6)10-11/h2-5H,1H3
Inchi Key
NZUUPXABAFLXFW-UHFFFAOYSA-N
Shipping Temperature
Normal
Purity
95%
Country of Origin
CN
Physical Form
Solid
Storage temp.
Sealed in dry, 2-8°C
Still not finding the right product?
Explore all of our products under 7-Chloro-2-methyl-2H-indazole
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
