Skip to Content
MilliporeSigma

Skip To

BDPH9BCC4EBE

Bide Pharmatech

Dipropyl malonate

Synonym(s):

dipropyl malonate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Linear Formula:
C9H16O4
CAS Number:
Molecular Weight:
188.22
MDL number:
Product BDPH9BCC4EBE is currently not available for purchase. Contact Technical Service


Inchi Code

1S/C9H16O4/c1-3-5-12-8(10)7-9(11)13-6-4-2/h3-7H2,1-2H3

Inchi Key

LWIWFCDNJNZEKB-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

98%

Storage temp.

Sealed in dry, room temperature

Country of Origin

CN

Physical Form

Colorless to pale yellow liquid



Still not finding the right product?

Explore all of our products under Dipropyl malonate




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library