Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Pack Size | SKU | Availability | Price |
|---|
About This Item
Product BDPH9BCC5C8A is currently not available for purchase. Contact Technical Service
Inchi Code
1S/C7H7NO2/c1-10-7-2-3-8-4-6(7)5-9/h2-5H,1H3
Inchi Key
QWUFIMUCFQUBOT-UHFFFAOYSA-N
Shipping Temperature
Normal
Purity
95%
Storage temp.
Inert atmosphere, 2-8C
Country of Origin
CN
Physical Form
Solid
Still not finding the right product?
Explore all of our products under 4-Methoxynicotinaldehyde
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
