Skip to Content
MilliporeSigma

Skip To

BDPH9BCCC2CD

Bide Pharmatech

5-Methylnicotinamide

Synonym(s):

5-methylnicotinamide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Linear Formula:
C7H8O1N2
CAS Number:
Molecular Weight:
136.15
MDL number:
Product BDPH9BCCC2CD is currently not available for purchase. Contact Technical Service


Inchi Code

1S/C7H8N2O/c1-5-2-6(7(8)10)4-9-3-5/h2-4H,1H3,(H2,8,10)

Inchi Key

BCCUXBGEPLKSEX-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

95%

Storage temp.

Keep in dark place, sealed in dry, room temperature

Country of Origin

CN

Physical Form

Yellow to Brown Solid



Still not finding the right product?

Explore all of our products under 5-Methylnicotinamide



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library