Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Pack Size | SKU | Availability | Price |
|---|
About This Item
Product BDPH9BCCC8EE is currently not available for purchase. Contact Technical Service
Inchi Code
1S/C6H10N2O/c1-5(2)8-4-6(9)3-7-8/h3-5,9H,1-2H3
Inchi Key
GTVJTOUCOLLRGC-UHFFFAOYSA-N
Shipping Temperature
Normal
Purity
97%
Storage temp.
Inert atmosphere, 2-8C
Country of Origin
CN
Physical Form
Solid
Still not finding the right product?
Explore all of our products under 1-Isopropyl-1H-pyrazol-4-ol
hstatements
signalword
Warning
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
