Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Pack Size | SKU | Availability | Price |
|---|
About This Item
Product BDPH9BCCE2DA is currently not available for purchase. Contact Technical Service
Inchi Code
1S/C10H9NO3/c12-9(13)6-11-5-7-3-1-2-4-8(7)10(11)14/h1-4H,5-6H2,(H,12,13)
Inchi Key
FPVHLXPCDHFPPN-UHFFFAOYSA-N
Shipping Temperature
Normal
Purity
98%
Storage temp.
Sealed in dry, 2-8C
Country of Origin
CN
Physical Form
Solid
Still not finding the right product?
Explore all of our products under 2-(1-Oxoisoindolin-2-yl)acetic acid
hstatements
signalword
Warning
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
