Skip to Content
MilliporeSigma

Skip To

BDPH9BCD2CD8

Bide Pharmatech

2-(1-Hydroxyethyl)-3,5-dimethylphenol

Synonym(s):

2-(1-hydroxyethyl)-3,5-dimethylphenol

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

Product BDPH9BCD2CD8 is currently not available for purchase. Contact Technical Service


Inchi Code

1S/C10H14O2/c1-6-4-7(2)10(8(3)11)9(12)5-6/h4-5,8,11-12H,1-3H3

Inchi Key

HXCGWXDSKDKHGO-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

95%

Storage temp.

Keep in dark place, sealed in dry, room temperature

Country of Origin

CN

Physical Form

Solid



Still not finding the right product?



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library