Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Pack Size | SKU | Availability | Price |
|---|
About This Item
Linear Formula:
C10H11O2N1Cl1Br1
CAS Number:
Molecular Weight:
292.56
MDL number:
Product BDPH9BCD2D2F is currently not available for purchase. Contact Technical Service
Inchi Code
1S/C10H11BrClNO2/c1-10(2,3)15-9(14)8-6(11)4-5-7(12)13-8/h4-5H,1-3H3
Inchi Key
JKGXLKXQUDHRLL-UHFFFAOYSA-N
Shipping Temperature
Normal
Purity
97%
Storage temp.
Inert atmosphere, 2-8C
Country of Origin
CN
Physical Form
White to Yellow Solid
Still not finding the right product?
Explore all of our products under tert-Butyl 3-bromo-6-chloropicolinate
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
