Skip to Content
MilliporeSigma

Skip To

BDPH9BCD2DAA

Bide Pharmatech

TRV-7019

Synonym(s):

(4-iodophenyl)methyl 6-methoxypyridine-2-carboxylate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Linear Formula:
C14H12O3N1I1
CAS Number:
Molecular Weight:
369.16
MDL number:
Product BDPH9BCD2DAA is currently not available for purchase. Contact Technical Service


Inchi Code

1S/C14H12INO3/c1-18-13-4-2-3-12(16-13)14(17)19-9-10-5-7-11(15)8-6-10/h2-8H,9H2,1H3

Inchi Key

AVKHPDWARYICID-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

98%

Storage temp.

Keep in dark place, sealed in dry, 2-8C

Country of Origin

CN

Physical Form

Solid



Still not finding the right product?

Explore all of our products under TRV-7019



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library