Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Pack Size | SKU | Availability | Price |
|---|
About This Item
Product BDPH9BCD4E80 is currently not available for purchase. Contact Technical Service
Inchi Code
1S/C12H16N2O4/c1-3-17-11(15)7-5-10(14)8(6-9(7)13)12(16)18-4-2/h5-6H,3-4,13-14H2,1-2H3
Inchi Key
KMKCBOQRRWZRII-UHFFFAOYSA-N
Shipping Temperature
Normal
Purity
95%
Storage temp.
Keep in dark place, inert atmosphere, room temperature
Country of Origin
CN
Physical Form
Solid
Still not finding the right product?
Explore all of our products under Diethyl 2,5-diaminoterephthalate
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
