Skip to Content
MilliporeSigma

Skip To

BDPH9BCDBDB2

Bide Pharmatech

2-Ethyl-4-methylphenol

Synonym(s):

2-ethyl-4-methylphenol

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Linear Formula:
C9H12O1
CAS Number:
Molecular Weight:
136.19
MDL number:
Product BDPH9BCDBDB2 is currently not available for purchase. Contact Technical Service


Inchi Code

1S/C9H12O/c1-3-8-6-7(2)4-5-9(8)10/h4-6,10H,3H2,1-2H3

Inchi Key

AVVVXUXMKWPKAJ-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

95%

Storage temp.

Inert atmosphere, room temperature

Country of Origin

CN

Physical Form

Liquid



Still not finding the right product?

Explore all of our products under 2-Ethyl-4-methylphenol




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library