Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Pack Size | SKU | Availability | Price |
|---|
About This Item
Product BDPH9BCDF4EB is currently not available for purchase. Contact Technical Service
Inchi Code
1S/C17H25BrSi/c1-13(2)19(14(3)4,15(5)6)12-11-16-7-9-17(18)10-8-16/h7-10,13-15H,1-6H3
Inchi Key
PTRSRXSVQYRPGE-UHFFFAOYSA-N
Shipping Temperature
Normal
Purity
98%
Storage temp.
Sealed in dry, room temperature
Country of Origin
CN
Physical Form
Liquid
Still not finding the right product?
Explore all of our products under ((4-Bromophenyl)ethynyl)triisopropylsilane
signalword
Warning
hstatements
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
