Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Pack Size | SKU | Availability | Price |
|---|
About This Item
Product BDPH9BCDF80F is currently not available for purchase. Contact Technical Service
Inchi Code
1S/C8H9FO2/c1-10-7-5-3-4-6(9)8(7)11-2/h3-5H,1-2H3
Inchi Key
MNBADDMXGOBTIC-UHFFFAOYSA-N
Shipping Temperature
Cold-chain transportation
Purity
95%
Storage temp.
Sealed in dry, room temperature
Country of Origin
CN
Physical Form
colorless Liquid
Still not finding the right product?
Explore all of our products under 1-Fluoro-2,3-dimethoxybenzene
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
