Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Pack Size | SKU | Availability | Price |
|---|
About This Item
Product BDPH9C02112E is currently not available for purchase. Contact Technical Service
Inchi Code
1S/C14H20O9/c1-6-11(20-7(2)15)12(21-8(3)16)13(22-9(4)17)14(19-6)23-10(5)18/h6,11-14H,1-5H3/t6-,11-,12+,13+,14-/m0/s1
Inchi Key
QZQMGQQOGJIDKJ-FWKODXMFSA-N
Shipping Temperature
Normal
Purity
98%
Storage temp.
Inert atmosphere, 2-8C
Country of Origin
CN
Physical Form
Solid or semi-solid or liquid
Still not finding the right product?
Explore all of our products under (2S,3R,4R,5S,6S)-6-Methyltetrahydro-2H-pyran-2,3,4,5-tetrayl tetraacetate
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.