Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Pack Size | SKU | Availability | Price |
|---|
About This Item
Product BDPH9C021DAD is currently not available for purchase. Contact Technical Service
Inchi Code
1S/C8H16O/c1-8(2)4-3-7(5-8)6-9/h7,9H,3-6H2,1-2H3
Inchi Key
LSZXVUJJTONWHR-UHFFFAOYSA-N
Shipping Temperature
Normal
Purity
95%
Storage temp.
Sealed in dry, room temperature
Country of Origin
CN
Physical Form
Liquid
Still not finding the right product?
Explore all of our products under (3,3-Dimethylcyclopentyl)methanol
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.