Skip to Content
MilliporeSigma

Skip To

BDPH9C021F9D

Bide Pharmatech

Certepetide

Synonym(s):

2,2'-((3S,6R,11R,14S,20S,23S,31aS)-23-(4-aminobutyl)-14-(3-guanidinopropyl)-1,4,12,15,18,21,24-heptahydroxy-6-(hydroxy(imino)methyl)-11-((1-hydroxyethylidene)amino)-27-oxo-6,7,10,11,14,17,20,23,26,27,29,30,31,31a-tetradecahydro-3H-pyrrolo[2,1-j][1,2]dithia[5,8,11,14,17,20,23,26]octaazacyclononacosine-3,20-diyl)diacetic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Linear Formula:
C37H60O14N14S2
CAS Number:
Molecular Weight:
989.1
MDL number:
Product BDPH9C021F9D is currently not available for purchase. Contact Technical Service


Inchi Code

1S/C37H60N14O14S2/c1-18(52)45-24-17-67-66-16-23(30(39)59)50-34(63)22(13-29(57)58)49-36(65)25-8-5-11-51(25)27(54)15-44-32(61)19(6-2-3-9-38)47-33(62)21(12-28(55)56)46-26(53)14-43-31(60)20(48-35(24)64)7-4-10-42-37(40)41/h19-25H,2-17,38H2,1H3,(H2,39,59)(H,43,60)(H,44,61)(H,45,52)(H,46,53)(H,47,62)(H,48,64)(H,49,65)(H,50,63)(H,55,56)(H,57,58)(H4,40,41,42)/t19-,20-,21-,22-,23-,24-,25-/m0/s1

Inchi Key

YPHPUVJQSYACEN-HUVRVWIJSA-N

Shipping Temperature

Cold-chain transportation

Purity

98%

Storage temp.

Keep in dark place, inert atmosphere, store in freezer, under -20C

Country of Origin

CN

Physical Form

Powder or crystals



Still not finding the right product?

Explore all of our products under Certepetide



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library