Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Pack Size | SKU | Availability | Price |
|---|
About This Item
Product BDPH9C021FFE is currently not available for purchase. Contact Technical Service
Inchi Code
1S/C8H4F3N/c9-6-1-2-7(10)8(11)5(6)3-4-12/h1-2H,3H2
Inchi Key
PQIFHBOOYBTJLD-UHFFFAOYSA-N
Shipping Temperature
Normal
Purity
97%
Storage temp.
Sealed in dry, 2-8C
Country of Origin
CN
Physical Form
Colorless to yellow to brown liquid
Still not finding the right product?
Explore all of our products under 2-(2,3,6-Trifluorophenyl)acetonitrile
hstatements
signalword
Warning
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
