Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Pack Size | SKU | Availability | Price |
|---|
About This Item
Product BL3H160C19DF is currently not available for purchase. Contact Technical Service
Inchi Code
1S/C8H9NO3/c9-7(8(11)12)5-2-1-3-6(10)4-5/h1-4,7,10H,9H2,(H,11,12)
Inchi Key
DQLYTFPAEVJTFM-UHFFFAOYSA-N
Shipping Temperature
Normal
Purity
98%
Storage temp.
Keep in dark place, sealed in dry, room temperature
Country of Origin
CN
Physical Form
Solid
Still not finding the right product?
Explore all of our products under 2-Amino-2-(3-hydroxyphenyl)acetic acid
hstatements
signalword
Warning
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
