MilliporeSigma
Skip To
469947
Tris(dimethylamido)aluminum(III)
Synonym(s):
Hexakis(dimethylamido)dialuminum, Tris(dimethylamino)alane dimer
Sign In to View Organizational & Contract Pricing.
About This Item
Linear Formula:
Al(N(CH3)2)3
Molecular Weight:
159.21
NACRES:
NA.23
PubChem Substance ID:
UNSPSC Code:
12352103
MDL number:
Quality Level
form
solid
reaction suitability
core: aluminum
mp
82-84 °C (lit.)
density
0.865 g/mL at 25 °C (lit.)
SMILES string
CN(C)[Al](N(C)C)N(C)C.CN(C)[Al](N(C)C)N(C)C
InChI
1S/6C2H6N.2Al/c6*1-3-2;;/h6*1-2H3;;/q6*-1;2*+3
InChI key
JGZUJELGSMSOID-UHFFFAOYSA-N
Compare Similar Items
View Full Comparison
Show Differences
1 of 1
This Item
663301669008668990
form
solid
form
liquid
form
liquid
form
liquid
density
0.865 g/mL at 25 °C (lit.)
density
0.752 g/mL at 25 °C (lit.)
density
0.947 g/mL at 25 °C (lit.)
density
1.252 g/mL at 25 °C (lit.)
mp
82-84 °C (lit.)
mp
15 °C (lit.)
mp
-
mp
-
Quality Level
100
Quality Level
100
Quality Level
100
Quality Level
100
reaction suitability
core: aluminum
reaction suitability
core: aluminum
reaction suitability
core: titanium
reaction suitability
core: tantalum
pictograms
FlameCorrosion
signalword
Danger
hcodes
supp_hazards
Storage Class
4.3 - Hazardous materials which set free flammable gases upon contact with water
wgk
WGK 3
flash_point_f
70.0 °F - closed cup
flash_point_c
21.1 °C - closed cup
ppe
Eyeshields, Faceshields, Gloves, type P3 (EN 143) respirator cartridges
Hazard Classifications
Skin Corr. 1B - Water-react. 1
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Visit the Document Library
Articles
Nanomaterials are considered a route to the innovations required for large-scale implementation of renewable energy technologies in society to make our life sustainable.
High Purity Metalorganic Precursors for CPV Device Fabrication
Tris (dimethylamido) aluminum (III): An overlooked atomic layer deposition precursor.
Buttera SC, et al.
Journal of Vacuum Science & Technology. A, Vacuum, Surfaces, And Films, 35(1), 01B128-01B128 (2017)
Global Trade Item Number
SKUGTIN
469947-10G04061833598610
Questions
Reviews
No rating value
Active Filters