MilliporeSigma
Get 22% off for Pi Day through March 31.Save Now
Skip To
EISCO-STDRD1
Eisco Labs
Eisco Labs Heavy Duty Rectangular Retort Base & Rod Set
Rectangular Retort Base & Rod Set - Base: 8"x5" - Rod: 23.6"L (60cm) x 12mm Dia. - Heavy Duty, Corrosion Resistant Steel
Shipped & Sold By Eisco Labs
A Trusted Marketplace Seller
Synonym(s):
Rectangular Retort Stand, Heavy Base with Rod, Laboratory Clamp Set, Physics Retort Setup, Heavy-Duty Support Stand
Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
Pack SizeSKUAvailabilityPrice
1 EA
Available to ship onApril 03, 2026
$34.50
$34.50
Available to ship onApril 03, 2026Details
Marketplace Support
Need help or have questions about this product?
Contact Seller
manufacturer/tradename
Eisco Labs
Related Categories
General description
  • HIGH QUALITY - Rod and base set consisting of a rectangular retort base made of corrosion-resistant, powder-coated pressed sheet metal steel and a rod made of stainless steel
  • STABILITY - Total weight of 3.75 lbs. The 8" long by 5" wide base weighs 2.6 lbs and is fitted with rubber feet, providing superb stability for experiments & setups - The included rod measures 23.6" (60cm) tall, is 12mm in diameter, and weighs 1.15 lbs
  • TAPPED HOLE AND THREADED ROD - Base features a tapped hole, which fits the included rod, but can accommodate other rods with a 10mm x 1.5mm metric thread and includes rubber sides to avoid surface scratching
  • GREAT FOR CLASSROOM & HOME LABORATORIES - The durable and sturdy construction of this set makes it an excellent choice for science classrooms, household laboratories & basic professional laboratory setups
  • MANUFACTURED BY EISCO LABS - Eisco Labs proudly manufactures its own products using state-of-the-art equipment, and following the highest standards of quality control. Each product is carefully designed by a team of laboratory professionals and experienced engineers with quality and durability in mind
2 Piece Laboratory Set - Rectangular Retort Base, 8"x5" & Stainless Steel Rod, 23.6"
Lab stands are used in every area of science - especially chemistry, for holding and manipulating an experimental apparatus. If you want to create a home laboratory, a retort base & rod are essential for any experiment setup.
The retort base is made of corrosion resistant, powder coated sheet metal steel, and features a tapped hole, suitable for connecting the included rod, or any other rods with a 10mm x 1.5mm metric thread. Base weighs 2.6 lbs, measures 8" long, 5" wide and approximately 2" in height, and is fitted with no-slip rubber feet providing superb stability for most experiments & setups. Included stainless steel rod weighs 1.15 lbs, measures 23.6" (60cm) tall, 12mm in diameter and features a 10x1.5mm metric thread. The durable and sturdy construction of this set makes it an excellent choice for science classrooms, household laboratories & basic professional laboratories.
Eisco Labs proudly manufactures its own products using state-of-the-art equipment, and following the highest standards of quality control.
Each product is carefully designed by a team of laboratory professionals and experienced engineers with quality and durability in mind. They are manufactured with only the highest grade, raw materials and according to international quality standards to ensure that they can withstand use in even the toughest laboratory environments.
Choose from one of the most recent versions:
Certificates of Analysis (COA)