Transition Metal Catalysts
Transition metals make excellent catalysts as they have incompletely filled d-orbitals that allow them to both donate and accept electrons easily from other molecules. Some of the early transition metal catalyzed reactions are still being used today. Despite their long history in catalysis, the discovery of new transition metal catalysts and making catalytic processes more efficient still remains an active area of research.
In addition, with the development of supporting ligands, they have evolved as a useful tool for various synthetic and non-synthetic transformations. Some examples of the transformations using transition metal catalysts include Stille, Buchwald-Hartwig, Negishi, Heck, Miyaura-Suzuki, and Sonogashira reactions.
No matter what your organic or organometallic catalysis requirement is, we have precisely the transition metal catalyst that you need.
Products
- Page 1
- Page 2
- Page 3
- Page 4
- Page 5
- Page 6
- Page 7
- Page 8
- Page 9
- Page 10
- Page 11
- Page 12
- Page 13
- Page 14
- Page 15
- Page 16
- Page 17
- Page 18
- Page 19
- Page 20
- Page 21
- Page 22
- Page 23
- Page 24
- Page 25
- Page 26
- Page 27
- Page 28
- Page 29
- Page 30
- Page 31
- Page 32
- Page 33
- Page 34
- Page 35
- Page 36
- Page 37
- Page 38
- Page 39
- Page 40
- Page 41
- Page 42
- Page 43
- Page 44
- Page 45
- Page 46
- Page 47
- Page 48
- Page 49
- Page 50
- Page 51
- Page 52
- Page 53
- Page 54
- Page 55
- Page 56
- Page 57
- Page 58
- Page 59
Related Resources
- Manual: C–H Functionalization Reaction
Learn about common C-H functionalization reactions so you can intuitively predict where and how to apply them and further explore the field with our new guide.
- Guide: Cross-Coupling Reaction
Cross-coupling reaction guide offers optimal conditions and catalyst systems for efficient chemical synthesis.
Vanadium Catalysts
The use of vanadium as a catalyst is the second largest application for vanadium after its use as an additive to improve steel production. A vanadium catalyst can effectively activate peroxides and selectively oxidize substrates like bromides, sulfides, and alkenes. These catalysts effectively transfer oxygen atoms to a substrate that is used for obtaining valuable oxidized molecules on a large-scale reaction with a high degree of selectivity. Additionally, vanadium catalysts are efficient catalysts for olefin polymerization. Vanadium oxides can be applied in the emission standards for vehicles and the desulfurization of crude oils. Moreover, the use of ecological oxidants, e.g., hydrogen and alkyl hydroperoxide, significantly increases the potential application of vanadium catalysts at an industrial level.
Iron Catalysts
Iron and iron compounds are widely used as reagents or catalysts. For example, ferric chloride and bromide have long been used as Lewis acid iron catalysts in the classic electrophilic aromatic substitution reactions. Iron complexes with organic ligands are of particular interest and can serve as environmentally friendly Fe catalysts for a host of transformations. Illustrating this point is the very useful role that iron catalysis plays in the timely study of ammonia-borane dehydrogenation process.
Cobalt Catalysts
Being both economical and ecological, cobalt catalysts have attracted intense interest for cross-coupling reactions. Cobalt catalysts are highly active reagents, extensively applied in the efficient and selective synthesis of pharmaceuticals, natural products, and new materials. These catalysts show a higher reactivity for various carbon-carbon bond formation reactions. Cobalt salts as catalysts show good functional group tolerance, high chemo-selectivity, and require mild reaction conditions in comparison to palladium and nickel, the most commonly used catalysts for metal-catalyzed cross-coupling.
Nickel Catalysts
Nickel catalysts play a central role in many synthetic transformations ranging from carbon-carbon cross coupling reactions to the reduction of electron-rich carbon bonds with Raney nickel. These nickel catalysts span a range of oxidation states: Nickel (0), nickel (II), nickel (III) and nickel (IV). The Ni catalysts available for immediate purchase are aluminum nickel (Al Ni) alloys, ammonium nickel hydrates, Ni COD, Ni halides (chlorides, bromides, fluorides and iodides), Ni cyclopentadienyls, nickel metal, nickel acac, and Raney Nickel― products of W.R. Grace and Company.
Copper Catalysts
Copper catalysts are useful for milder reaction conditions and show excellent yields, however the chemical reactions are slow and require high temperatures. Among transition-metal mediated reactions to form carbon-carbon bonds and carbon-heteroatom bonds, copper catalysts are used in Ullmann reactions, Diels-Alder reactions, ring expansions, Castro-Stevens coupling, the Kharasch-Sosnovsky reaction, and a notable variant of the Huisgen 1,3-dipolar cycloaddition utilizing a Cu(I) catalyst developed independently by Meldal and Sharpless. We provide efficient copper catalysts and pre-catalysts as well as copper-containing Metal Organic Framework (MOF) components for all your copper catalysis needs.
Zinc Catalysts
Zinc catalysts find wide applicability in synthetic chemistry and organic synthesis. A zinc chloride catalyst, acting as a moderate-strength Lewis acid, can catalyze the Fischer Indole synthesis to convert aryl hydrazones to indoles, and the Friedel-Crafts Acylation to produce monoacylated products from arenes and acyl chlorides. In addition to ZnCl2, a zinc oxide catalyst can be useful in a variety of catalytic conversions. We also offer additional zinc catalysts, such as various zinc halides, that catalyze stereospecific and regioselective reactions. In addition to the catalytic properties, our zinc compounds also find applications in material science as chemiluminescent quantum dots and nanomaterials. Our zinc compounds may also be used as starting materials in the preparation of organozinc reagents used in Negishi coupling.
Zirconium Catalysts
The zirconium-catalyzed asymmetric carboalumination (ZACA) reaction, developed by Nobel laureate Ei-ichi Negishi, is perhaps one of the best-known examples of use of zirconium catalyst. The ZACA reaction provides a means for chiral functionalization of alkenes with organoaluminum agents, catalyzed by a chiral bis(indenyl)zirconium catalyst. Another notable zirconium catalyst is zirconium dioxide or zirconia. The list of applications of zirconia catalyst in heterogeneous catalysis is rapidly growing. Some of the applications include: decomposition of nitric oxide, reduction of carboxylic acid to aldehydes, selective dehydration of secondary alcohols to terminal alkenes, and hydrogenation of carbon monoxide to isobutane.
Ruthenium Catalysts
Ruthenium Catalysts
Selective oxidative transformation of various functional groups with environment friendly and easily accessible oxidants can be readily achieved with a proper ruthenium catalyst. Ruthenium catalysts can be very powerful tool in synthetic chemistry for selective catalysis of oxidative transformations such as asymmetric epoxidation of alkenes, generation of dioxygen species, dihydroxylation of olefins, and oxidative dehydrogenation of alcohols.
Ruthenium catalysts are also widely employed in metathesis reactions, with Grubbs′ catalysts being the most well-known in the field of olefin metathesis. The wide popularity of Grubb′s catalysts can be explained by their high tolerance of various functional groups, and their high stability in air and a plethora of solvents.
Rhodium Catalysts
The rhodium catalyst is a suitable promoter, used to activate carbon-hydrogen(C-H) bonds, and has emerged as a challenging and attractive tool for catalysis. Rhodium catalysis finds increasing interest in the catalytic dehydrogenative cross-coupling, allowing elegant C-C bond construction. Although palladium has been the metal of choice for most examples, Rh catalysts can also be suitable promoters for this activation. Additionally, rhodium provides access to important couplings, such as aryl-aryl, aryl-alkene, and alkene-alkene, as viable routes to valuable organic frameworks.
Palladium Catalysts
The ability to fine tune the reaction conditions (temperature, solvents, ligands, bases and other additives) of palladium catalysts makes palladium catalysis an extremely versatile tool in organic chemical synthesis. Furthermore, palladium catalysts have a very high tolerance of various functional groups and are often able to provide excellent stereo- and regio- specificity, which helps to avoid the need for protecting groups. It forms a highly versatile group of catalysts, known for use in carbon bond forming reactions (primarily C-C, C-O, C-N and C-F), such as Heck coupling, Suzuki coupling, Stille coupling, Hiyama coupling, Sonogashira coupling, Negishi coupling, and Buchwald-Hartwig amination, among others.
In heterogeneous catalysis, palladium catalysts, such as the Lindlar catalyst (or Lindlar′s Palladium), are highly efficient at facilitating selective hydrogenation. This includes the conversion of triple bonds to cis-double bonds, monohydrogenation of polyolefins, and hydrogenation of azides to amines.
We welcome you to review our extensive offering of highly versatile homogeneous and heterogeneous palladium catalysts. For even greater convenience in purification and post-reaction cleanup, we have also included a selection of supported Pd catalysts, as well as a full line of recyclable and immobilized Pd Encat® catalysts that are suitable for various bond-formation and hydrogenation/reduction reactions.
Silver Catalysts
Our portfolio also offers a wide variety of high-quality silver catalysts for transition-metal catalysis in organic synthesis. Silver catalysts are commonly used due to the high oxidation power and high oxidation potential of silver complexes. Additionally, they also serve as silver activators and enhance the electronegativity of other catalysts, such as gold. Organic and inorganic synthesis benefit from the stoichiometric oxidation potential of silver compounds. Homogeneous silver-catalyzed organic transformations highlight the unique redox chemistry of silver, capable of catalyzing reactions with high stereo- and regioselectivity. Silver catalysts efficiently mediate both intermolecular as well as intramolecular bond formations. Heterogeneous processes involving silver catalysis include NOx reduction and catalytic oxidation of carbon monoxide (CO) to carbon dioxide (CO2). Silver(I) salts are also used in several silver-catalyzed nucleophilic addition reactions and organic transformations.
Platinum Catalysts
We provide efficient platinum catalysts, e.g., platinum dioxide, also called Adams′ catalyst, for the hydrogenation of various functional groups and dehydrogenation reactions in organic synthesis. Platinum black, the active Pt catalyst, is formed during the reaction. Utilizing platinum catalysis on alkynes results in syn-addition, forming a cis-alkene. Two of the most important transformations using platinum catalysts include the hydrogenation of nitro compounds to amines and ketones to alcohols. Notably, reductions of alkenes can also be performed with Adams′ catalyst in the presence of nitro groups without reducing the nitro group. Platinum catalysts are preferred over palladium catalysts to minimize hydrogenolysis when reducing nitro compounds to amines. This Pt catalyst is also used for the hydrogenolysis of phenyl phosphate esters, a reaction that does not occur with palladium catalysts.
Gold Catalysts
Prior to the 1980s, gold was regarded as having little catalytic activity. Advancements, spearheaded by F. Dean Toste (University of California, Berkeley) and others, have propelled gold into the forefront of transition metal catalysis. In particular, phosphine-ligated gold(I) complexes have recently emerged as powerful C–C bond forming catalysts capable of performing a variety of reactions under mild conditions. The list of useful C–C bond construction methods includes cyclopropanations, enyne isomerizations, Rautenstrauch rearrangements, ene reactions, and ring expansions. Typically, the catalyst system relies on a phosphine gold(I) chloride complex in combination with a silver salt co-catalyst to generate the active species in situ.